Human Recombinant SARS-CoV-2 (COVID-19) Spike S1 Protein (from HEK293 cells)
Catalog #: 77132-010
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:43740568
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Conjugation:Unconjugated
- Size:50 µg
- Storage conditions:Short term storageL: 4 °C, long term storage: −20 °C, Handling advice: After opening, prepare aliquots and store at −20 °C. Avoid freeze/thaw cycles. Centrifuge lyophilized vial before opening and reconstitution. For maximum product recovery after thawing, centrifuge the vial before opening the cap. Use/Stability: Stable for at least 6 months after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
- Biological activity:Binds to the human ACE2.
- Protein synonyms:2019-nCoV Spike Protein S1 (RBD)Spike Receptor Binding Domain
- Protein/peptide name:Spike
- Purity:≥95% (SDS-PAGE)
- Molecular weight:~60 kDa (SDS-PAGE)
- Sequence:Receptor-binding domain (RBD) of SARS-CoV-2 Spike protein S1 (aa 319-541) is fused to the N-terminus of the Fc region of human IgG1
- Concentration:1 mg/ml after reconstitution
- Formulation:Lyophilized. Lyophilized. Contains PBS.
- Shipping temperature:Room temperature
Specifications
About this item
Customers Who Bought This Also Bought