Order Entry
ContactUsLinkComponent
SARS-CoV-2 (COVID-19) Recombinant Spike Protein (from bacteria)
SARS-CoV-2 (COVID-19) Recombinant Spike Protein (from bacteria)
Catalog #: 77129-690
Supplier:  Prosci
New Product
CAS Number:  
SARS-CoV-2 (COVID-19) Recombinant Spike Protein (from bacteria)
Catalog #: 77129-690
Supplier:  Prosci
New Product
CAS Number:  

Specifications

  • Gene ID:
    43740568
  • Packaging:
    Vial
  • Protein/peptide type:
    Recombinant
  • Source:
    Bacteria
  • Species:
    SARS-CoV-2
  • Conjugation:
    Unconjugated
  • Size:
    0.1 mg
  • Storage conditions:
    Store at −70 °C. Avoid repeated freeze-thaw cycles.
  • Protein synonyms:
    Spike Protein SARS-CoV2
    SARS-CoV-2 Spike Protein
    SARS-CoV2-S Protein
    S Protein SARS-CoV-2
    SARS-CoV-2 S Protein
    SARS-CoV2 Spike Protein
  • Protein/peptide name:
    Spike
  • Purity:
    Amylose resin (0.85)
  • Molecular weight:
    Mono-Isotopic Mass: 185, 299.77 daltons Average Mass: 185, 417.39 daltons
  • Concentration:
    Batch dependent
  • Formulation:
    Liquid. 50 mM Tris-HCl pH 7.5, 270 mM Sucrose, 150 mM NaCl, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35
  • Shipping temperature:
    Dry ice

Specifications

About this item

Native Sequence: MKIEEGKLVIWINGDKGYNGLAEVGKKFEKDTGIKVTVEHPDKLEEKFPQVAATGDGPDIIFWAHDRFGGYAQSGLLAEITPDKAFQDKLYPFTWDAVRYNGKLIAYPIAVEALSLIYNKDLLPNPPKTWEEIPALDKELKAKGKSALMFNLQEPYFTWPLIAADGGYAFKYENGKYDIKDVGVDNAGAKAGLTFLVDLIKNKHMNADTDYSIAEAAFNKGETAMTINGPWAWSNIDTSKVNYGVTVLPTFKGQPSKPFVGVLSAGINAASPNKELAKEFLENYLLTDEGLEAVNKDKPLGAVALKSYEEELVKDPRIAATMENAQKGEIMPNIPQMSAFWYAVRTAVINAASGRQTVDEALKDAQTNSSSNNNNNNNNNNLGDDDDKVPEFLEVLFQGPLGSMFVFLVLLPLVSSQCVNLTTRTQLPPAYTNSFTRGVYYPDKVFRSSVLHSTQDLFLPFFSNVTWFHAIHVSGTNGTKRFDNPVLPFNDGVYFASTEKSNIIRGWIFGTTLDSKTQSLLIVNNATNVVIKVCEFQFCNDPFLGVYYHKNNKSWMESEFRVYSSANNCTFEYVSQPFLMDLEGKQGNFKNLREFVFKNIDGYFKIYSKHTPINLVRDLPQGFSALEPLVDLPIGINITRFQTLLALHRSYLTPGDSSSGWTAGAAAYYVGYLQPRTFLLKYNENGTITDAVDCALDPLSETKCTLKSFTVEKGIYQTSNFRVQPTESIVRFPNITNLCPFGEVFNATRFASVYAWNRKRISNCVADYSVLYNSASFSTFKCYGVSPTKLNDLCFTNVYADSFVIRGDEVRQIAPGQTGKIADYNYKLPDDFTGCVIAWNSNNLDSKVGGNYNYLYRLFRKSNLKPFERDISTEIYQAGSTPCNGVEGFNCYFPLQSYGFQPTNGVGYQPYRVVVLSFELLHAPATVCGPKKSTNLVKNKCVNFNFNGLTGTGVLTESNKKFLPFQQFGRDIADTTDAVRDPQTLEILDITPCSFGGVSVITPGTNTSNQVAVLYQDVNCTEVPVAIHADQLTPTWRVYSTGSNVFQTRAGCLIGAEHVNNSYECDIPIGAGICASYQTQTNSPRRARSVASQSIIAYTMSLGAENSVAYSNNSIAIPTNFTISVTTEILPVSMTKTSVDCTMYICGDSTECSNLLLQYGSFCTQLNRALTGIAVEQDKNTQEVFAQVKQIYKTPPIKDFGGFNFSQILPDPSKPSKRSFIEDLLFNKVTLADAGFIKQYGDCLGDIAARDLICAQKFNGLTVLPPLLTDEMIAQYTSALLAGTITSGWTFGAGAALQIPFAMQMAYRFNGIGVTQNVLYENQKLIANQFNSAIGKIQDSLSSTASALGKLQDVVNQNAQALNTLVKQLSSNFGAISSVLNDILSRLDKVEAEVQIDRLITGRLQSLQTYVTQQLIRAAEIRASANLAATKMSECVLGQSKRVDFCGKGYHLMSFPQSAPHGVVFLHVTYVPAQEKNFTTAPAICHDGKAHFPREGVFVSNGTHWFVTQRNFYEPQIITTDNTFVSGNCDVVIGIVNNTVYDPLQPELDSFKEELDKYFKNHTSPDVDLGDISGINASVVNIQKEIDRLNEVAKNLNESLIDLQELGKYEQYIKWPWYIWLGFIAGLIAIVMVTIMLCCMTSCCSCLKGCCSCGSCCKFDEDDSEPVLKGVKLHYT Amino acids M1 – T1273 (end). Residue M404 of the fusion protein is equivalent to M1 of the native enzyme. The MBP tag is located at residues 1 – 367. Protease Cleavage: PreScission (LEVLFQGP) residues 393 – 400

Customers Who Bought This Also Bought