Order Entry
ContactUsLinkComponent
HCoV-OC43 Recombinant Human Coronavirus Nucleocapsid (229E) Protein (from bacteria)
HCoV-OC43 Recombinant Human Coronavirus Nucleocapsid (229E) Protein (from bacteria)
Catalog #: 77133-844
Supplier:  Prosci
New Product
CAS Number:  
HCoV-OC43 Recombinant Human Coronavirus Nucleocapsid (229E) Protein (from bacteria)
Catalog #: 77133-844
Supplier:  Prosci
New Product
CAS Number:  

Specifications

  • Gene ID:
    918763
  • Packaging:
    Vial
  • Protein/peptide type:
    Recombinant
  • Source:
    Bacteria
  • Species:
    HCoV-OC43
  • Conjugation:
    Unconjugated
  • Size:
    0.1 mg
  • Storage conditions:
    Store at −70 °C. Avoid repeated freeze-thaw cycles.
  • Protein synonyms:
    Human CoV 229E Nucleocapsid Protein
    Human CoV 229E N Protein
  • Protein/peptide name:
    Nucleocapsid
  • Purity:
    Amylose Resin (0.85)
  • Molecular weight:
    Mono-Isotopic Mass: 87, 651.64 daltons Average Mass: 87, 705.68 daltons
  • Concentration:
    Batch dependent
  • Formulation:
    Liquid. 50 mM Tris-HCl pH 7.5, 270 mM Sucrose, 150 mM NaCl, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35
  • Shipping temperature:
    Dry ice

Specifications

About this item

Native Sequence: MKIEEGKLVIWINGDKGYNGLAEVGKKFEKDTGIKVTVEHPDKLEEKFPQVAATGDGPDIIFWAHDRFGGYAQSGLLAEITPDKAFQDKLYPFTWDAVRYNGKLIAYPIAVEALSLIYNKDLLPNPPKTWEEIPALDKELKAKGKSALMFNLQEPYFTWPLIAADGGYAFKYENGKYDIKDVGVDNAGAKAGLTFLVDLIKNKHMNADTDYSIAEAAFNKGETAMTINGPWAWSNIDTSKVNYGVTVLPTFKGQPSKPFVGVLSAGINAASPNKELAKEFLENYLLTDEGLEAVNKDKPLGAVALKSYEEELVKDPRIAATMENAQKGEIMPNIPQMSAFWYAVRTAVINAASGRQTVDEALKDAQTNSSSNNNNNNNNNNLGDDDDKVPEFLEVLFQGPLGSMATVKWADASEPQRGRQGRIPYSLYSPLLVDSEQPWKVIPRNLVPINKKDKNKLIGYWNVQKRFRTRKGKRVDLSPKLHFYYLGTGPHKDAKFRERVEGVVWVAVDGAKTEPTGYGVRRKNSEPEIPHFNQKLPNGVTVVEEPDSRAPSRSQSRSQSRGRGESKPQSRNPSSDRNHNSQDDIMKAVAAALKSLGFDKPQEKDKKSAKTGTPKPSRNQSPASSQTSAKSLARSQSSETKEQKHEMQKPRWKRQPNDDVTSNVTQCFGPRDLDHNFGSAGVVANGVKAKGYPQFAELVPSTAAMLFDSHIVSKESGNTVVLTFTTRVTVPKDHPHLGKFLEELNAFTREMQQHPLLNPSALEFNPSQTSPATAEPVRDEVSIETDIIDEVN Amino acids M1 – N389 (end). Residue M404 of the fusion protein is equivalent to M1 of the native enzyme. The MBP tag is located at residues 1 – 367. Protease Cleavage: PreScission (LEVLFQGP) residues 393 - 400

Customers Who Bought This Also Bought