SARS Recombinant SARS Coronavirus Envelope (HSZ-Cc) Protein (from bacteria)
Catalog #: 77126-786
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:1489671
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:Bacteria
- Species:SARS
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:SARS-CoV-HSZ-Cc Envelope ProteinSARS-CoV-HSZ-Cc-E ProteinE Protein SARS-CoV-HSZ-CcEnvelope Protein SARS-CoV-HSZ-Cc
- Protein/peptide name:Envelope
- Purity:Amylose resin (0.75)
- Molecular weight:Mono-Isotopic Mass: 52, 566.88 daltons Average Mass: 52, 599.92 daltons
- Sequence:MKIEEGKLVIWINGDKGYNGLAEVGKKFEKDTGIKVTVEHPDKLEEKFPQVAATGDGPDIIFWAHDRFGGYAQSGLLAEITPDKAFQDKLYPFTWDAVRYNGKLIAYPIAVEALSLIYNKDLLPNPPKTWEEIPALDKELKAKGKSALMFNLQEPYFTWPLIAADGGYAFKYENGKYDIKDVGVDNAGAKAGLTFLVDLIKNKHMNADTDYSIAEAAFNKGETAMTINGPWAWSNIDTSKVNYGVTVLPTFKGQPSKPFVGVLSAGINAASPNKELAKEFLENYLLTDEGLEAVNKDKPLGAVALKSYEEELVKDPRIAATMENAQKGEIMPNIPQMSAFWYAVRTAVINAASGRQTVDEALKDAQTNSSSNNNNNNNNNNLGDDDDKVPEFLEVLFQGPLGSMYSFVSEETGTLIVNSVLLFLAFVVFLLVTLAILTALRLCAYCCNIVNVSLVKPTVYVYSRVKNLNSSEGVPDLLV Amino acids M1 – V76 (end). Residue M404 of the fusion protein is equivalent to M1 of the native enzyme. The MBP tag is located at residues 1 – 367. Protease Cleavage: PreScission (LEVLFQGP) residues 393 – 400
- Concentration:Batch dependent
- Formulation:Liquid. 50 mM Tris-HCl pH 7.5, 270 mM Sucrose, 150 mM NaCl, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35
- Shipping temperature:Dry ice
Specifications
About this item
Customers Who Bought This Also Bought