Mouse Recombinant CD155/PVR Protein (from HEK293 cells)
77129-576
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:0.05 mg
- Storage conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:52118
- Protein synonyms:TAGE4|PVS|FLJ25946|CD155|NECL5|PVR|HVED
- Protein/peptide name:Pvr
- Purity:>95% as determined by reduced SDS-PAGE
- Molecular weight:61.8 kDa
- Sequence:Asp 29 - Leu 348
- Formulation:Lyophilized. Tris with Glycine, Arginine and NaCl, pH 7.5
- Shipping temperature:RT
- Tested applications:ELISA,WB
- Cat. no.:77129-576