Human Recombinant Mucin-1/MUC-1 (1036-1155) Protein (from HEK293 cells)
Catalog #: 77125-956
Supplier: Prosci
New Product
CAS Number:
Specifications
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:Mucin 1EpisialinCA 15-3PEMTPUMMUC-1/XCD227MUC-1/SECKL-6EMAMUC1MAM6PEMH23AGMUC1/ZD
- Protein/peptide name:Mucin-1/MUC-1 (1036-1155)
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:7.2 kDa (partial α chain) and 8.2 kDa (partial β chain)
- Sequence:Leu 1036 - Gly 1155
- Formulation:Lyophilized. PBS, pH 7.4
- Shipping temperature:Room temperature
- Tested applications:WB
Specifications
About this item
Customers Who Bought This Also Bought