Cynomolgus monkey Recombinant Latent TGF-beta 1 (C33S) Protein (from HEK293 cells)
Catalog # 77124-746
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Size:0.05 mg
- Storage conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:CED|TGF-beta-1|TGFB|DPD1|Latent TGF-beta 1|Latent TGFB1|LAP|TGFB1
- Protein/peptide name:Latent TGF-beta 1 (C33S)
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:43.1 kDa
- Sequence:Leu 30 - Ser 390 (C33S)
- Formulation:Lyophilized. PBS, pH 7.4
- Shipping temperature:RT
- Tested applications:ELISA,WB
- Cat. no.:77124-746