Cynomolgus monkey Recombinant IL-1 Rrp2/IL-1 R6 Protein (from HEK293 cells)
Catalog # 77127-036
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Size:0.1 mg
- Storage conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:IL1RL2|IL-1RL2|IL-36R|IL-1Rrp2|IL1Rrp2|IL-1 R6|IL1RRP2|IL-1 Rrp2
- Protein/peptide name:IL-1 Rrp2/IL-1 R6
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:37.9 kDa
- Sequence:Asp 20 - Arg 335
- Formulation:Lyophilized. PBS, pH 7.4
- Shipping temperature:RT
- Tested applications:WB
- Cat. no.:77127-036