Anti-SCN4B Rabbit Polyclonal Antibody
Catalog # 76753-282
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:Sodium channel, type IV, beta
- Antigen symbol:SCN4B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:399548
- Antigen synonyms:sodium channel subunit beta-4|voltage gated sodium channel beta 4 subunit|sodium channel accessory subunit|beta polypeptide|type IV|Gm1471|sodium channel
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant protein of mouse SCN4B.
- Purification:Affinity purification
- Cat. no.:76753-282
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SCN4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat