Anti-KLF4 Rabbit Polyclonal Antibody
Catalog # 76756-568
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:Kruppel-like factor 4 (gut)
- Antigen symbol:KLF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:9314
- Antigen synonyms:Epithelial zinc finger protein EZF|GKLF|KLF4 Antibody: EZF|EZF|Krueppel-like factor 4
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 180-479 of human KLF4 (NP_004226.3).
- Purification:Affinity purification
- Cat. no.:76756-568
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: KLF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat