Anti-TMF1 Rabbit Polyclonal Antibody
76751-328
: Prosci
- Antibody type:Primary
- Antigen name:TATA element modulatory factor 1
- Antigen symbol:TMF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:7110
- Antigen synonyms:androgen receptor-associated protein of 160 kDa|ARA16TMF|androgen receptor coactivator 160 kDa protein|TATA element modulatory factor
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 900-1000 of human TMF1 (NP_009045.2).
- Purification:Affinity purification
- Cat. no.:76751-328
- Packaging:Vial
Type: Primary
Antigen: TMF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human