Anti-ABCA2 Rabbit Polyclonal Antibody
76761-044
: Prosci
- Antibody type:Primary
- Antigen name:ATP-binding cassette, sub-family A (ABC1), member 2
- Antigen symbol:ABCA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:20
- Antigen synonyms:sub-family A (ABC1)|ABC2|OTTHUMP00000064733|member 2|KIAA1062|ABCA2|MGC129761|ATP-binding cassette|sub-family A
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 2307-2466 of human ABCA2 (NP_997698.1).
- Purification:Affinity purification
- Cat. no.:76761-044
- Packaging:Vial
Type: Primary
Antigen: ABCA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse