Anti-PPM1K Rabbit Polyclonal Antibody
76756-182
: Prosci
- Antibody type:Primary
- Antigen name:Protein phosphatase, Mg2+/Mn2+ dependent 1K
- Antigen symbol:PPM1K
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:152926
- Antigen synonyms:PP2C-kappa|PP2Cm|BDP|PP2C-type mitochondrial phosphoprotein phosphatase|MSUDMV|protein phosphatase 1K|PP2Ckappa|mitochondrial|PTMP|protein phosphatase 2C kappa|branched-chain I+/--ketoacid dehydrogenase phosphatase|branched-chain alpha-ketoacid dehydrogenase phosphatase|PP2C domain-containing protein phosphatase 1K|UG0882E07
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 183-372 of human PPM1K (NP_689755.3).
- Purification:Affinity purification
- Cat. no.:76756-182
- Packaging:Vial
Type: Primary
Antigen: PPM1K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat