Anti-OR2S2 Rabbit Polyclonal Antibody
Catalog # 76763-858
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:Olfactory receptor family 2 subfamily S member 2 (gene/pseudogene)
- Antigen symbol:OR2S2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:56656
- Antigen synonyms:subfamily S|OR37A|olfactory receptor 2S2|olfactory receptor OR9-3|member 2|OST715|family 2|olfactory receptor
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 200-300 of human OR2S2 (NP_063950.2).
- Purification:Affinity purification
- Cat. no.:76763-858
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: OR2S2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human