Anti-KIF7 Rabbit Polyclonal Antibody
50 µl, Vial
Catalog # 76758-832
Supplier: Prosci
Supplier #: 16-121
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type: Primary
Antigen: KIF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Size:50 µl
- Packaging:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Modification:None
- Purification:Affinity purification
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Concentration:Batch dependent
- Shipping temperature:20
- Antigen name:Kinesin family member 7
- Cat. No.:76758-832
- Reactivity:Human
- Gene ID:374654
- Antigen synonyms:AGBK|ACLS|JBTS12|HLS2|EQYK340|UNQ34kinesin-like protein KIF7
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1044-1343 of human KIF7 (NP_940927.2).
- Conjugation:Unconjugated
- Antigen symbol:KIF7
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3



