Anti-THNSL1 Rabbit Polyclonal Antibody
76758-622
: Prosci
- Antibody type:Primary
- Antigen name:Threonine synthase-like 1
- Antigen symbol:THNSL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:79896
- Antigen synonyms:THNSL1|Threonine synthase-like 1|TSH1
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 534-743 of human THNSL1 (NP_079114.3).
- Purification:Affinity purification
- Cat. no.:76758-622
- Packaging:Vial
Type: Primary
Antigen: THNSL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat