Anti-ZNF549 Rabbit Polyclonal Antibody
Catalog # 76754-500
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:Zinc finger protein 549
- Antigen symbol:ZNF549
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:256051
- Antigen synonyms:zinc finger protein 549
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ZNF549 (NP_001186224.1).
- Purification:Affinity purification
- Cat. no.:76754-500
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: ZNF549
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human