Anti-DNAJC19 Rabbit Polyclonal Antibody
76763-880
: Prosci
- Antibody type:Primary
- Antigen name:Mitochondrial import inner membrane translocase subunit TIM14
- Antigen symbol:DNAJC19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:131118
- Antigen synonyms:Mitochondrial import inner membrane translocase subunit TIM14|TIM14|TIMM14|DNAJC19|DnaJ homolog subfamily C member 19
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-116 of human DNAJC19 (NP_660304.1).
- Purification:Affinity purification
- Cat. no.:76763-880
- Packaging:Vial
Type: Primary
Antigen: DNAJC19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat