Anti-MRPL22 Rabbit Polyclonal Antibody
Catalog # 76751-454
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:Mitochondrial ribosomal protein L22
- Antigen symbol:MRPL22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:29093
- Antigen synonyms:MRP-L25|MRP-L22|L22mt|mitochondrial|RPML25|39S ribosomal protein L25|L25mt|mitochondrial large ribosomal subunit protein uL22m|39S ribosomal protein L22|HSPC158
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 97-206 of human MRPL22 (NP_054899.2).
- Purification:Affinity purification
- Cat. no.:76751-454
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: MRPL22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat