Anti-ARPC1A Rabbit Polyclonal Antibody
Catalog # 76756-604
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:Actin related protein 2/3 complex, subunit 1A, 41 kDa
- Antigen symbol:ARPC1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:10552
- Antigen synonyms:actin-related protein 2/3 complex subunit 1A|OTTHUMP00000206034|subunit 1A|Arc40|actin binding protein (Schizosaccharomyces pombe sop2-like)|41kDa|SOP2Hs|actin related protein 2/3 complex|SOP2-like protein|SOP2L|ARPC1A
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 171-350 of human ARPC1A (NP_006400.2).
- Purification:Affinity purification
- Cat. no.:76756-604
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: ARPC1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse