Anti-NPEPL1 Rabbit Polyclonal Antibody
Catalog # 76753-882
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:Aminopeptidase like 1
- Antigen symbol:NPEPL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:79716
- Antigen synonyms:probable aminopeptidase NPEPL1|bA261P9.2
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 374-523 of human NPEPL1 (NP_078939.3).
- Purification:Affinity purification
- Cat. no.:76753-882
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: NPEPL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse