Anti-ACOT2 Rabbit Polyclonal Antibody
76759-326
: Prosci
- Antibody type:Primary
- Antigen name:Acyl-CoA thioesterase 2
- Antigen symbol:ACOT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:10965
- Antigen synonyms:ACOT2|CTE1A|ZAP128|PTE2A|PTE2|MTE1|CTE-IA|Mte1
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 222-483 of human ACOT2 (NP_006812.3).
- Purification:Affinity purification
- Cat. no.:76759-326
- Packaging:Vial
Type: Primary
Antigen: ACOT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat