Anti-MAP2K6 Rabbit Polyclonal Antibody
Catalog # 76753-202
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase 6
- Antigen symbol:MAP2K6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:5608
- Antigen synonyms:PRKMK6|Mapk kinase 6|MKK6|MAPKK6|MAP kinase kinase 6|SAPK kinase 3|SAPKK-3|MAP2K6|MEK6|SKK3|MAPK/ERK kinase 6|SAPKK3|MEK 6|MAPKK 6
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-334 of human MAP2K6 (NP_002749.2).
- Purification:Affinity purification
- Cat. no.:76753-202
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: MAP2K6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat