Anti-DBNL Rabbit Polyclonal Antibody
76748-930
: Prosci
- Antibody type:Primary
- Antigen name:Drebrin-like
- Antigen symbol:DBNL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:28988
- Antigen synonyms:SH3P7|PP5423|HIP-55|drebrin F|CMAP|src homology 3 domain-containing protein HIP-55|HIP-55 protein
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 131-430 of human DBNL (NP_001014436.1).
- Purification:Affinity purification
- Cat. no.:76748-930
- Packaging:Vial
Type: Primary
Antigen: DBNL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat