Anti-MECOM Rabbit Polyclonal Antibody
Catalog # 76753-538
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:MDS1 and EVI1 complex locus
- Antigen symbol:MECOM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:2122
- Antigen synonyms:AML1-EVI-1|MGC163392|MDS1|MDS1-EVI1|EVI-1|EVI1|PRDM3
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human MECOM (NP_004982.2).
- Purification:Affinity purification
- Cat. no.:76753-538
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: MECOM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse