Anti-TIGIT Llama Recombinant Monoclonal Antibody [clone: 2C6]
Catalog # 77121-998
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:T-cell immunoreceptor with Ig and ITIM domains
- Antigen symbol:TIGIT
- Clone:2C6
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Llama
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Size:0.02 mg
- Form:Liquid
- Gene ID:2016332
- Antigen synonyms:TIGIT Antibody: T-cell immunoreceptor with Ig and ITIM domains|WUCAM|VSTM3|VSIG9
- Modification:None
- Storage buffer:TIGIT antibody is supplied in PBS.
- Storage temperature:TIGIT antibody should be stored in working aliquots at −20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:Dry Ice
- Immunogen:TIGIT antibody was raised against a recombinant protein corresponding to amino acids 22-141 of human TIGIT.
- Purification:TIGIT antibody is affinity chromatography purified via Nickel column. Antibody is supplied as a his-tagged purified rotein. It also contains a myc-tag for detection.
- Cat. no.:77121-998
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: TIGIT
Clonality: Recombinant Monoclonal
Clone: 2C6
Conjugation: Unconjugated
Epitope:
Host: Llama
Isotype: sdAb
Reactivity: Human