Anti-PDCD1 Llama Recombinant Monoclonal Antibody [clone: F4-2D7]
Catalog # 77124-668
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:programmed cell death 1
- Antigen symbol:PDCD1
- Clone:F4-2D7
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Llama
- Reactivity:Human
- Size:0.02 mg
- Form:Liquid
- Gene ID:5133
- Antigen synonyms:PD-1|hPD-1|hPD-l|Programmed cell death protein 1|PDCD-1|PD-1 Antibody: PD1|CD279|hSLE1|Protein PD-1|PDCD1|PD1|SLEB2
- Modification:None
- Storage buffer:PD-1 antibody is supplied in PBS.
- Storage temperature:PD-1 antibody should be stored in working aliquots at −20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:Dry Ice
- Immunogen:PD-1 antibody was raised against a recombinant protein corresponding to amino acids 25-167 of human PD-1.
- Purification:PD-1 antibody is affinity chromatography purified via Nickel column. Antibody is supplied as a his-tagged purified rotein. It also contains a myc-tag for detection.
- Cat. no.:77124-668
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PDCD1
Clonality: Recombinant Monoclonal
Clone: F4-2D7
Conjugation: Unconjugated
Epitope:
Host: Llama
Isotype: sdAb
Reactivity: Human