Anti-Il2 Rat Monoclonal Antibody [clone: [JES6-1A12]]
Catalog # 77125-198
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:interleukin 2
- Antigen symbol:Il2
- Clonality:Monoclonal
- Clone:[JES6-1A12]
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Size:0.5 mg
- Form:Liquid
- Gene ID:16183
- Antigen synonyms:KHF|killer cell helper factor|MAF-C I|Interleukin-2|EDF|TCGF|thymocyte differentiation factor|eosinophil differentiation factor|T cell growth factor|macrophage-activating factor for cytotoxicity I|TDF
- Modification:None
- Storage buffer:Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added.
- Storage temperature:Store vial at 2...8 °C.
- Concentration:0.5 mg/ml
- Shipping temperature:20
- Immunogen:E. coli-expressed mouse IL-2.
- Cat. no.:77125-198
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: Il2
Clonality: Monoclonal
Clone: [JES6-1A12]
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse