Anti-CR1 Mouse Monoclonal Antibody [clone: [E11]] (FITC (Fluorescein Isothiocyanate))
Catalog # 77125-230
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:complement component (3b/4b) receptor 1 (Knops blood group)
- Antigen symbol:CR1
- Clonality:Monoclonal
- Clone:[E11]
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Size:100 tests
- Form:Liquid
- Gene ID:1378
- Antigen synonyms:C4bR|complement receptor type 1|C3bR|CR1|immune adherence receptor
- Modification:None
- Storage buffer:Supplied in PBS/NaN3.
- Storage temperature:Store vial at 2...8 °C.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Acute monocytic leukemia cells and normal blood monocytes.
- Cat. no.:77125-230
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CR1
Clonality: Monoclonal
Clone: [E11]
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rhesus Monkey, Cynomolgus Monkey, Chimpanzee, Baboon