Anti-Cd28 Hamster Monoclonal Antibody [clone: [PV-1]]
Catalog # 77131-736
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:CD28 antigen
- Antigen symbol:Cd28
- Clonality:Monoclonal
- Clone:[PV-1]
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Hamster
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Size:0.5 mg
- Form:Liquid
- Gene ID:12487
- Antigen synonyms:Tp44|T44
- Modification:None
- Storage buffer:Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added.
- Storage temperature:Store vial at 2...8 °C.
- Concentration:0.5 mg/ml
- Shipping temperature:20
- Immunogen:C57BL/6N mouse T cell lymphoma EL4 cell line.
- Cat. no.:77131-736
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: Cd28
Clonality: Monoclonal
Clone: [PV-1]
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse