Anti-COL4A1 Mouse Monoclonal Antibody [clone: [2F11]]
76762-260
: Prosci
- Antibody type:Primary
- Antigen name:collagen type IV alpha 1 chain
- Antigen symbol:COL4A1
- Clonality:Monoclonal
- Clone:[2F11]
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:1282;1284;1285;1286;1287;1288
- Antigen synonyms:COLIV
- Modification:None
- Storage buffer:Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added.
- Storage temperature:Store vial at 2...8 °C.
- Concentration:0.5 mg/ml
- Shipping temperature:20
- Immunogen:Native human type IV collagen.
- Cat. no.:76762-260
- Packaging:Vial
Type: Primary
Antigen: COL4A1
Clonality: Monoclonal
Clone: [2F11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human