Anti-PEX5 Goat Polyclonal Antibody
Catalog # 77119-754
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:peroxisomal biogenesis factor 5
- Antigen symbol:PEX5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:5830
- Antigen synonyms:peroxisomal biogenesis factor 5|peroxin-5|OTTHUMP00000238353|OTTHUMP00000238194|FLJ50721|peroxisomal C-terminal targeting signal import receptor|PTS1-BP|OTTHUMP00000238195|OTTHUMP00000238354|OTTHUMP00000238192|PEX5|OTTHUMP00000238356|OTTHUMP00000238193|PTS1R|peroxisomal targeting signal receptor 1|peroxisomal targeting signal 1 receptor|peroxisomal targeting signal 1 (SKL type) receptor|PTS1 receptor|peroxisome receptor 1|FLJ50634|FLJ51948|PXR1|peroxisomal targeting signal import receptor
- Modification:None
- Storage buffer:Supplied at 0.5 mg/ml in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
Aliquot and store at -20°C. Minimize freezing and thawing. - Storage temperature:Aliquot and store at −20 °C. Minimize freezing and thawing.
- Concentration:500 µg/ml
- Shipping temperature:20
- Immunogen:The immunogen for this antibody is: C-QHTASDFVAKVDDPK.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:77119-754
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PEX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human