Anti-HIF3A Goat Polyclonal Antibody
Catalog # 77127-500
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:hypoxia inducible factor 3, alpha subunit
- Antigen symbol:HIF3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Size:0.1 mg
- Form:Liquid
- Gene ID:64344
- Antigen synonyms:hypoxia inducible factor 3|PASD7|HIF3A|HIF-3A4|inhibitory PAS domain protein|alpha subunit|HIF-3A2|IPAS|hypoxia-inducible factor-3 alpha 4|bHLHe17|MOP7|HIF-3A
- Modification:None
- Storage buffer:Supplied at 0.5 mg/ml in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
Aliquot and store at -20°C. Minimize freezing and thawing. - Storage temperature:Aliquot and store at −20 °C. Minimize freezing and thawing.
- Concentration:500 µg/ml
- Shipping temperature:20
- Immunogen:The immunogen for this antibody is: QDHRSTTELRKEKS-C.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:77127-500
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: HIF3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human