Anti-AP2A1 Goat Polyclonal Antibody
Catalog # 77130-862
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:adaptor-related protein complex 2, alpha 1 subunit
- Antigen symbol:AP2A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:160
- Antigen synonyms:AP-2 complex subunit alpha-1|alpha 1|adapter-related protein complex 2 alpha-1 subunit|alpha-adaptin A|CLAPA1|AP2A1|plasma membrane adaptor HA2/AP2 adaptin alpha A subunit|alpha A|AP2-ALPHA|adaptin|alpha 1 subunit|clathrin assembly protein complex 2 alpha-A large chain|ADTAA|large|adaptor-related protein complex 2|adaptor protein complex AP-2 subunit alpha-1|clathrin-associated/assembly/adaptor protein|alpha1-adaptin|100 kDa coated vesicle protein A
- Modification:None
- Storage buffer:Supplied at 0.5 mg/ml in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
Aliquot and store at -20°C. Minimize freezing and thawing. - Storage temperature:Aliquot and store at −20 °C. Minimize freezing and thawing.
- Concentration:500 µg/ml
- Shipping temperature:20
- Immunogen:The immunogen for this antibody is: C-ELEPPAPESPMALLADPAPAAD.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:77130-862
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: AP2A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse