Anti-SAV1 Rabbit Polyclonal Antibody
76758-182
: Prosci
- Antibody type:Primary
- Antigen name:salvador homolog 1 (Drosophila)
- Antigen symbol:SAV1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:60485
- Antigen synonyms:WW45|WW domain-containing|WW45 protein|salvador homolog 1 (Drosophila)|1700040G09Rik|SAV|SAV1|WWP4
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human SAV1 (NP_068590.1).
- Purification:Affinity purification.
- Cat. no.:76758-182
- Packaging:Vial
Type: Primary
Antigen: SAV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human