Anti-MRPS6 Rabbit Polyclonal Antibody
Catalog # 76754-610
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:mitochondrial ribosomal protein S6
- Antigen symbol:MRPS6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:64968
- Antigen synonyms:MRP-S6|S6mt|RPMS6|mitochondrial|C21orf101|28S ribosomal protein S6|mitochondrial small ribosomal subunit protein bS6m
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human MRPS6 (NP_115865.1).
- Purification:Affinity purification.
- Cat. no.:76754-610
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: MRPS6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat