Anti-PAM16 Rabbit Polyclonal Antibody
Catalog # 76760-358
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:presequence translocase associated motor 16
- Antigen symbol:PAM16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:51025
- Antigen synonyms:MAGMAS|magmas-like protein|presequence translocase associated motor 16 homolog|CGI-136|mitochondria-associated granulocyte macrophage CSF-signaling molecule|mitochondrial import inner membrane translocase subunit TIM16|TIM16|TIMM16|mitochondria associated protein involved in granulocyte macrophage colony stimulating factor signal transduction|SMDMDM
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 21-125 of human PAM16 (NP_057153.8).
- Purification:Affinity purification.
- Cat. no.:76760-358
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PAM16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse