Anti-PRAP1 Rabbit Polyclonal Antibody
Catalog # 76751-458
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:proline rich acidic protein 1
- Antigen symbol:PRAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:118471
- Antigen synonyms:epididymis secretory sperm binding protein|proline-rich acidic protein 1|PRO1195|epididymis tissue protein Li 178|UPA
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-151 of human PRAP1 (NP_660203.3).
- Purification:Affinity purification.
- Cat. no.:76751-458
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PRAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human