Anti-CGB7 Rabbit Polyclonal Antibody
Catalog # 76760-634
Supplier: Prosci
Specifications
- Antibody type:Primary
- Antigen name:chorionic gonadotropin subunit beta 7
- Antigen symbol:CGB7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:94027
- Antigen synonyms:beta polypeptide 7|choriogonadotropin subunit beta 7|chorionic gonadotropin beta 7 subunit|CG-beta-a|chorionic gonadotropin beta subunit 7|CGB6|chorionic gonadotropin
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 21-165 of human CGB7 (NP_149133.1).
- Purification:Affinity purification.
- Cat. no.:76760-634
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CGB7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat