Order Entry
Canada
Orders LinkContactUsLinkComponent
LMX1B Peptide

LMX1B Peptide

Vial, 0.1 mg
Catalog # 77121-410
Supplier:   Prosci
Supplier #:  42-043P
LMX1B Peptide
Catalog # 77121-410
Supplier:  Prosci
CAS Number:  
LMX1B Peptide

Vial, 0.1 mg

CAS Number:  
Price
$477.16
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    0.1 mg
  • Cat. No.:
    77121-410
  • Protein/peptide type:
    Synthetic
  • Protein/peptide name:
    LMX1B
  • Storage conditions:
    Store at −20 °C. When reconstituted minimize freezing and thawing.
  • Gene ID:
    4010
  • Formulation:
    Lyophilized. LMX1B peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
  • Protein synonyms:
    NPS1|LIM homeobox transcription factor 1-beta|LMX1.2
  • Conjugation:
    Unconjugated
  • Tested applications:
    Blocking
  • Shipping temperature:
    RT

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.