SALL1 Peptide
Vial, 0.1 mg
Catalog # 77131-494
Supplier: Prosci
Supplier #: 42-194P
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Specifications
Specifications
- Pack type:Vial
- Size:0.1 mg
- Cat. No.:77131-494
- Protein/peptide type:Synthetic
- Protein/peptide name:SALL1
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Gene ID:6299
- Formulation:Lyophilized. SALL1 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Protein synonyms:HSAL1|Spalt-like transcription factor 1|HEL-S-89|ZNF794|Sal-1|Sal-like protein 1|TBS|SAL1|HSal1
- Conjugation:Unconjugated
- Tested applications:Blocking
- Shipping temperature:RT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



