Order Entry
Canada
ContactUsLinkComponent
Lifeasible Bacillus thuringiensis Recombinant Vip3A from (E. coli)
Lifeasible Bacillus thuringiensis Recombinant Vip3A from (E. coli)
Catalog # 76683-062
Supplier:  Creative Biomart
CAS Number:  
Lifeasible Bacillus thuringiensis Recombinant Vip3A from (E. coli)
Catalog # 76683-062
Supplier:  Creative Biomart
Supplier Number:  VIP3A-03B
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Insecticidal protein
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Bacillus thuringiensis
  • Size:
    1 mg
  • Storage conditions:
    –80 °C
  • Animal-free:
    Y
  • Protein/peptide name:
    Vip3A
  • Purity:
    50%
  • Molecular weight:
    60 - 70 kDa
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    –80 °C
  • Cat. no.:
    76683-062

Specifications

About this item

B. thuringiensis (Bt) is a gram-positive, soil-dwelling, spore-forming, rod-shaped bacteria. It is approximately 1 µm in width and 5 µm in length. It grows at body temperature and produces a diamond-shaped crystal from its crystal proteins (Cry proteins) and uses it to fend off insects, predators, and other pathogens.

Delivery: Dry ice.