Lifeasible Bacillus thuringiensis Recombinant Cry3B from (E. coli)
76683-096
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Insecticidal protein
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Bacillus thuringiensis
- Size:1 mg
- Storage conditions:–80 °C
- Animal-free:Y
- Protein/peptide name:Cry3B
- Purity:85%
- Molecular weight:70 - 90 kDa
- Concentration:1 mg/ml
- Shipping temperature:–80 °C
- Cat. no.:76683-096
Promotes colloidosmotic lysis by binding to the midgut epithelial cells of lepidopteran (Manduca sexta) larvae.
: Dry ice.