Order Entry
Canada
ContactUsLinkComponent
Lifeasible Bacillus thuringiensis Recombinant Cry1A.105 from (E. coli)
Lifeasible Bacillus thuringiensis Recombinant Cry1A.105 from (E. coli)
Catalog # 76683-090
Supplier:  Creative Biomart
CAS Number:  
Lifeasible Bacillus thuringiensis Recombinant Cry1A.105 from (E. coli)
Catalog # 76683-090
Supplier:  Creative Biomart
Supplier Number:  CRY1A105-09B
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Insecticidal protein
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Bacillus thuringiensis
  • Size:
    1 mg
  • Storage conditions:
    –80 °C
  • Animal-free:
    Y
  • Protein/peptide name:
    Cry1A.105
  • Purity:
    90%
  • Molecular weight:
    136.8 kDa
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    –80 °C
  • Cat. no.:
    76683-090

Specifications

About this item

Cry1A.105 is an engineered chimeric protein composed of domains I and II from Cry1Ab or Cry1Ac, domain III from Cry1F, and a C-terminal domain from Cry1Ac, and has insecticidal activity against a variety of pests. The Cry1A.105 protein is activated by the intestinal juice of the target pest to obtain an activated toxin fragment, which binds to the midgut epithelial cells of the pest to form holes, leading to intestinal paralysis and eventually death due to sepsis.

Sequence: MDNNPNINECIPYNCLSNPEVEVLGGERIETGYTPIDISLSLTQFLLSEFVPGAGFVLGLVDIIWGIFGPSQWDAFLVQIEQLINQRIEEFARNQAISRLEGLSNLYQIYAESFREWEADPTNPALREEMRIQFNDMNSALTTAIPLFAVQNYQVPLLSVYVQAANLHLSVLRDVSVFGQRWGFDAATINSRYNDLTRLIGNYTDHAVRWYNTGLERVWGPDSRDWIRYNQFRRELTLTVLDIVSLFPNYDSRTYPIRTVSQLTREIYTNPVLENFDGSFRGSAQGIEGSIRSPHLMDILNSITIYTDAHRGEYYWSGHQIMASPVGFSGPEFTFPLYGTMGNAAPQQRIVAQLGQGVYRTLSSTLYRRPFNIGINNQQLSVLDGTEFAYGTSSNLPSAVYRKSGTVDSLDEIPPQNNNVPPRQGFSHRLSHVSMFRSGFSNSSVSIIRAPMFSWIHRSAEFNNIIASDSITQIPLVKAHTLQSGTTVVRGPGFTGGDILRRTSGGPFAYTIVNINGQLPQRYRARIRYASTTNLRIYVTVAGERIFAGQFNKTMDTGDPLTFQSFSYATINTAFTFPMSQSSFTVGADTFSSGNEVYIDRFELIPVTATLEAEYNLERAQKAVNALFTSTNQLGLKTNVTDYHIDQVSNLVTYLSDEFCLDEKRELSEKVKHAKRLSDERNLLQDSNFKDINRQPERGWGGSTGITIQGGDDVFKENYVTLSGTFDECYPTYLYQKIDESKLKAFTRYQLRGYIEDSQDLEIYSIRYNAKHETVNVPGTGSLWPLSAQSPIGKCGEPNRCAPHLEWNPDLDCSCRDGEKCAHHSHHFSLDIDVGCTDLNEDLGVWVIFKIKTQDGHARLGNLEFLEEKPLVGEALARVKRAEKKWRDKREKLEWETNIVYKEAKESVDALFVNSQYDQLQADTNIAMIHAADKRVHSIREAYLPELSVIPGVNAAIFEELEGRIFTAFSLYDARNVIKNGDFNNGLSCWNVKGHVDVEEQNNQRSVLVVPEWEAEVSQEVRVCPGRGYILRVTAYKEGYGEGCVTIHEIENNTDELKFSNCVEEEIYPNNTVTCNDYTVNQEEYGGAYTSRNRGYNEAPSVPADYASVYEEKSYTDGRRENPCEFNRGYRDYTPLPVGYVTKELEYFPETDKVWIEIGETEGTFIVDSVELLLMEE

Delivery: Dry ice.