GRN Peptide
Catalogue # 77124-286
Fournisseur: Prosci
Numéro CAS:
Spécifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Synthetic
- Species:Human
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Gene ID:2896
- Protein synonyms:PGRN|GEP|Proepithelin|PCDGF|PEPI|GP88|Granulins|CLN11
- Protein/peptide name:GRN
- Formulation:Lyophilized. GRN peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:RT
- Tested applications:Blocking
- Cat. no.:77124-286