SARS-CoV-2 (COVID-19) Recombinant Delta Variant (B.1.617.2) Spike RBD (L452R, T478K) Protein (from Mammalian Cells)
Catalog #: 77128-826
Supplier: Prosci
New Product
CAS Number:
Specifications
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:Mammalian cells
- Species:SARS-CoV-2
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2...8 °C for one week. Store at −20...− 80 °C for twelve months from the date of receipt.
- Protein synonyms:Spike RBD ProteinRBD ProteinIN variant (B.1.617.2)
- Protein/peptide name:Spike RBD (L452R, T478K)
- Purity:>90% as determined by SDS-PAGE
- Sequence:RBD with mutations as L452R and T478K.
- Formulation:Lyophilized. Supplied as Lyophilized from PBS, pH 7.5. Please reconstitute the protein with ddH₂O to the concentration with 1 mg/ml.
- Shipping temperature:Room temperature
Specifications
About this item
Customers Who Bought This Also Bought