Cynomolgus, Rhesus macaque Recombinant CD27/TNFRSF7 Protein (from HEK293 cells)
Catalog # 77127-804
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkeyRhesus monkey
- Size:0.1 mg
- Storage conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:102139476
- Protein synonyms:Tp55|S152|T14|TNFRSF7|CD27
- Protein/peptide name:CD27
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:46.2 kDa
- Sequence:Ala 20 - Arg 191
- Formulation:Lyophilized. 50 mM Tris, 100 mM Glycine, pH 7.5.
- Shipping temperature:RT
- Tested applications:ELISA,WB
- Cat. no.:77127-804