Human Recombinant IL-1 Rrp2/IL-1 R6 Protein (from HEK293 cells)
Catalog #: 77129-374
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:8808
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:IL1RL2IL-1RL2IL-36RIL-1Rrp2IL1Rrp2IL-1 R6IL1RRP2IL-1 Rrp2
- Protein/peptide name:IL1RL2
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:62.4 kDa
- Sequence:Asp 20 - Arg 335
- Formulation:Lyophilized. Tris with Glycine, Arginine and NaCl, pH 7.5.
- Shipping temperature:Room temperature
- Tested applications:ELISA,WB
Specifications
About this item
Customers Who Bought This Also Bought