Human Recombinant GFR alpha-like Protein (from HEK293 cells)
Catalog # 77129-482
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.1 mg
- Storage conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:389400
- Protein synonyms:GFRAL|GRAL|GFR alpha like|GFR alpha-like
- Protein/peptide name:GFRAL
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:39.7 kDa
- Sequence:Ser 19 - Glu 351
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping temperature:RT
- Tested applications:WB
- Cat. no.:77129-482