Rhesus monkey Recombinant Rhesus macaque Complement Factor D/CFD Protein (from HEK293 cells)
Catalog #: 77128-536
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:102136571
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Rhesus monkey
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:PFDComplement factor DCFDDFAdipsin
- Protein/peptide name:CFD
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:51.0 kDa
- Sequence:Ile 26 - Ala 253
- Formulation:Lyophilized. Tris with Glycine, Arginine and NaCl, pH 7.5.
- Shipping temperature:Room temperature
- Tested applications:WB
Specifications
About this item
Customers Who Bought This Also Bought